Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:Galectin 1
- Antigen-Symbol:LGALS1
- Klonalität:Polyklonal
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Technische Daten
Über diesen Artikel
Polyclonal antibody for GALECTIN 1/LGALS1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. GALECTIN 1/LGALS1 information: Molecular Weight: 14716 MW; Subcellular Localization: Secreted, extracellular space, extracellular matrix ; Tissue Specificity: Expressed in placenta, maternal decidua and fetal membranes. Within placenta, expressed in trophoblasts, stromal cells, villous endothelium, syncytiotrophoblast apical membrane and villous stroma. Within fetal membranes, expressed in amnion, chorioamniotic mesenchyma and chorion (at protein level). Expressed in cardiac, smooth, and skeletal muscle, neurons, thymus, kidney and hematopoietic cells.
Type: Primary
Antigen: Galectin-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat