Anti-Angiopoietin-1 Polyclonal Antibody
Katalognr. BSBTPB9125
Anbieter: Boster Bio
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:Angiopoietin-1
- Klonalität:Polyklonal
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Technische Daten
Über diesen Artikel
Polyclonal antibody for ANGIOPOIETIN 1/ANGPT1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. ANGIOPOIETIN 1/ANGPT1 information: Molecular Weight: 57513 MW; Subcellular Localization: Secreted.
Type: Primary
Antigen: Angiopoietin-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human