Elemental analysis standards, PTP sample of oils, VHG
Katalognr. PROOVHG-VPTPMO-25G
Anbieter: LGC Standards PROMOCHEM
CAS-Nummer:
Technische Daten
- VE:25 g
- Bezeichnung:PTP sample of oils, VHG, 25 g
Technische Daten
Über diesen Artikel
The PTP has been specially designed for analytical laboratories that analyse new and used oils for metals, sulphur, viscosity and particle count.
- Monitor and improve measurements processes
- Demonstrate analytical competency
- Method and instrumentation validation
- 25 analytes
Certifications: ISO/IEC 17043 accredited.