Pipette tips, Sapphire
Codice catalogo GREI771255_960
Nome del fornitore: Greiner Bio-One
Specifiche
- Verpackt:Rack (96 tips/rack)
- Steril:-
- Volumen:10 µl
- Seq:02
- Spitzentyp:Graduated, low retention
- Spitzenfarbe:Transparent
- VE:960
Specifiche
Informazioni su questo articolo
These tips are manufactured from virgin materials. All tips are transparent, graduated, and allow precise and comfortable pipetting with maximal recovery.
- Graduations for perfect visual control of liquid
- Thin-walled top for optimal seal
- Free of detectable DNase, RNase, human DNA and non pyrogenic
- Available as low retention tips for even higher recovery