ContactUsLinkComponent
Reference standards, soil, BTEX - Loamy Sand 1, CRM
Reference standards, soil, BTEX - Loamy Sand 1, CRM
Catalog # PROORTC-CRM308
CAS Number:  
Reference standards, soil, BTEX - Loamy Sand 1, CRM
Catalog # PROORTC-CRM308
CAS Number:  
Specifications
  • VE:
    30 g
  • Bezeichnung:
    BTEX - Loamy Sand 1, (5 analytes)
  • Environmentally Preferable:
Specifications
About this item
Certifications: Traceable to NIST, SRM.