Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Levamisole Hydrochloride, Reference Standard | Avantor

Levamisole Hydrochloride, Reference Standard

Levamisole Hydrochloride, USP reference standard, Vial, 125 mg
Katalognr. 1359302.
Anbieter:   USP
Levamisole Hydrochloride, Reference Standard
Katalognr. 1359302.
Anbieter:  USP
CAS Nummer:  
Levamisole Hydrochloride, Reference Standard

Levamisole Hydrochloride, USP reference standard, Vial, 125 mg

CAS Nummer:  
Preis
SFr. 363.00
Einloggen für Verfügbarkeit
1 * 125 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Zertifizierung: US Pharmacopea Reference Standard

Technische Daten

Technische Daten

  • Packung:
    Vial
  • VE:
    125 mg
  • Normen:
    USP reference standard
  • Bezeichnung:
    Levamisole Hydrochloride
  • UN No.:
    2811
  • Cat. No.:
    1359302.
  • CAS Nr.:
    16595-80-5
  • Bezeichnung:
    Levamisole Hydrochloride
  • Formel:
    C₁₁H₁₃ClN₂S

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.