Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant DAN
  PRSI92-561
 :  
Mouse recombinant DAN
  PRSI92-561
 :  

 

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Spezies:
    Mouse
  • Tag sequence:
    C-6 His tag
  • Lagerbedingungen:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Proteinsynonym:
    Protein N03|suppression of tumorigenicity 1|neuroblastoma suppressor of tumorigenicity 1|NBL1|DAND1|DAN domain family member 1
  • Protein/Peptid-Name:
    DAN
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18.4 kD
  • Sequence:
    Ala17-Asp178
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

 

 

Differential screening-selected gene aberrative in neuroblastoma (DAN) is a member of the DAN family of secreted glycoproteins. DAN family antagonists are Characterised by a DAN domain that contains a cystine knot motif which is essential for binding to BMP ligands. Members of this family include DAN, gremlin, protein related to DAN and cerberus (PRDC), cerberus, sclerostin (SOST) and uterine sensitisation-associated gene 1 protein, and control diverse processes in growth, development and the cell cycle. It has also been reported that DAN family plays crucial role in early mouse embryo development by inhibiting the action of bone morphogenic proteins and modulating the action of transforming growth factor- beta superfamily members. DAN is Synthesised by small-to intermediate-sised DRG neurons and transported to the sensory nerve terminals in the skin or to the sensory nerve terminals in the dorsal horn. It has been reported that DAN is ubiquitously expressed in adult rat and human tissues. Morphological studies have revealed that, in adult rat, DAN mRNA is expressed ubiquitously in lung and brain, but not in liver.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.