Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant IL18 (from E. coli)
  PRSI92-246
 :  
Human recombinant IL18 (from E. coli)
  PRSI92-246
 :  

 

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Lagerbedingungen:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Proteinsynonym:
    IL-1g|IL-1 gamma|IL-1F4|MGC12320|Interferon gamma-inducing factor|Iboctadekin|GIF|Interleukin-1 gamma|IL1F4|IFN-gamma-inducing factor|Interleukin-18|IL-18
  • Protein/Peptid-Name:
    IL18
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18.2 kD
  • Sequence:
    Tyr37-Asp193
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Validierte Anwendungen:
    Bioassay

 

 

Interleukin-18 is a secreted protein and it belongs to the IL-1 family. IL-18 is a proinflammatory cytokine and produced by macrophages and other cells. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL-4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein (IL18BP) can specifically interact with this cytokine, and thus negatively regulate its biological activity. After stimulation with IL-18, natural killer (NK) cells and certain T cells release another important cytokine called interferon- gamma (IFN- gamma ) or type II interferon that plays an important role in activating the macrophages or other cells.

This recombinant protein can be used for biological assays. For research use only.