Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant MIF (from E. coli)
Mouse recombinant MIF (from E. coli)
Catalog # PRSI92-244
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant MIF (from E. coli)
Catalog # PRSI92-244
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Mouse
  • Tag sequence:
    C-6 His tag
  • Lagerbedingungen:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Proteinsynonym:
    Phenylpyruvate tautomerase|Glycosylation-inhibiting factor|Delayed early response protein 6|L-dopachrome tautomerase|Macrophage migration inhibitory factor|GIF|L-dopachrome isomerase|DER6
  • Protein/Peptid-Name:
    MIF
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    13.5 kD
  • Sequence:
    Pro2-Ala115
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Validierte Anwendungen:
    Bioassay

Specifications

About this item

Macrophage migration inhibitory factor(MIF) is a secreted protein and belongs to the MIF family. MIF is an important regulator of innate immunity. The circulating MIF binds to CD74 on other immune cells to trigger an acute immune response. Hence MIF is classified as an inflammatory cytokine. Furthermore glucocorticoids also stimulate white blood cells to release MIF and hence MIF partially counter acts the inhibitory effects that glucocorticoids have on the immune system. Finally trauma activates the anterior pituitary gland to release MIF.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.