Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant UBE2J2 (from E. coli)
Human recombinant UBE2J2 (from E. coli)
Catalog # PRSI91-996
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant UBE2J2 (from E. coli)
Catalog # PRSI91-996
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Tag sequence:
    N-GST tag
  • Lagerbedingungen:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Proteinsynonym:
    UBE2J2|NCUBE2|Non-Canonical Ubiquitin-Conjugating Enzyme 2|NCUBE-2|Ubiquitin-Conjugating Enzyme E2 J2
  • Protein/Peptid-Name:
    UBE2J2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    37.96 kD
  • Sequence:
    Glu124-Arg224
  • Zusammensetzung:
    Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM DTT, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Validierte Anwendungen:
    Bioassay

Specifications

About this item

Ubiquitin-Conjugating Enzyme E2 J2 (UBE2J2) belongs to the ubiquitin-conjugating enzyme family. UBE2J2 is involved in the ubiquitiantion. UBE2J2 located in the membrane of the endoplasmic reticulum, catalyzes the covalent attachment of ubiquitin to other proteins. UBE2J2 may play a important role in the selective degradation of misfolded membrane protein from the endoplasmic reticulum.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-GST tag