Human recombinant TNF receptor II (from cells)
Référence PRSI91-624
Fournisseur: ProSci Inc.
Numéro CAS:
Spécifications
- VE:50 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:Cells
- Spezies:Human
- Tag sequence:C-6 His tag
- Lagerbedingungen:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Proteinsynonym:CD120b|Tumor necrosis factor receptor superfamily member 1B|TNFRSF1B|p75|p80 TNF-alpha receptor|TNFR-II|TNF-R2|Tumor necrosis factor receptor type II|Tumor necrosis factor receptor 2|TNF-RII
- Protein/Peptid-Name:TNF Receptor II
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:19 kD
- Sequence:Lys288-Ser461
- Zusammensetzung:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Validierte Anwendungen:Bioassay
Spécifications
À propos de cet article
Tumor necrosis factor receptor superfamily member 1B(TNFRSF1B) is expressed by the gene TNFRSF1B. The soluble form is produced from the membrane form by proteolytic processing. It can bind to TRAF2, and interacts with BMX. It can act as the receptor with high affinity for TNFSF2/TNF-alpha and approximately 5-fold lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. This receptor mediates most of the metabolic effects of TNF-alpha.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag