Order Entry
Switzerland
ContactUsLinkComponent
 
Human Recombinant GRP (from E. coli)
  PRSI91-210
 :  
Human Recombinant GRP (from E. coli)
  PRSI91-210
 :  

 

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Tag sequence:
    N-6 His tag
  • Lagerbedingungen:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Proteinsynonym:
    LSM4|GRP|Glycine-Rich Protein|U6 snRNA-Associated Sm-like Protein LSm4
  • Protein/Peptid-Name:
    GRP
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.5 kD
  • Sequence:
    Met1-Gln139
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 1mM DTT, pH 8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Validierte Anwendungen:
    For Biological assays

 

 

U6 snRNA-associated Sm-like protein LSm4 (LSM4) is a member of the snRNP Sm proteins family. Sm-like proteins contain the Sm sequence motif and are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing. LSM4 forms a heteromer with a donut shape. The complexes are involved in various steps of RNA metabolism. LSM4 binds specifically to the 3-terminal U-tract of U6 snRNA. LSM4 contributes RNA protein interactions and structural changes which are essential during ribosomal subunit assembly.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag