ContactUsLinkComponent
Human recombinant GAS7 (from E. coli)
Catalog # PRSI91-205
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant GAS7 (from E. coli)
Catalog # PRSI91-205
Supplier:  ProSci Inc.
CAS Number:  
Specifications
  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Tag sequence:
    N-6 His tag
  • Lagerbedingungen:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Proteinsynonym:
    KIAA0394|GAS-7|GAS7|Growth Arrest-Specific Protein 7
  • Protein/Peptid-Name:
    GAS7
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    49.4 kD
  • Sequence:
    Met1-Ile412
  • Zusammensetzung:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 2mM DTT, 10% Glycerol, pH 8.8. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Validierte Anwendungen:
    Bioassay
Specifications
About this item
Growth Arrest-Specific Protein 7 (GAS7) is expressed primarily in terminalaly differentiated brain cells and predominantly in mature cerebellar Purkinje neurons. GAS7 may play a role in neuronal development by promoting maturation and morphological differentiation of cerebellar neurons. Inhibition of GAS7 production in terminally differentiating cultures of embryonic murine cerebullum impedes neurite outgrowth. The hyper-expression of GAS7 may play an major role in the initiation and development of huaman osteosarcoma.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag