Human recombinant CLIC5 (from E. coli)
50 µG
Katalognr. PRSI91-192
Anbieter: ProSci Inc.
Human recombinant CLIC5 (from <i>E. coli</i>)
Katalognr. PRSI91-192
Anbieter: ProSci Inc.
CAS Nummer:
Direkt zu
Product Details & Documents
Chloride Intracellular Channel Protein 5 (CLIC5) is a single-pass membrane protein which belongs to the chloride channel CLIC family. It contains one GST C-terminal domain. Chloride intracellular channels are involved in chloride ion transport within various subcellular compartments. CLIC5 can insert into membranes and form selective ion channels regulated by actin that may transport chloride ions. It may play a role in the regulation of transepithelial ion absorption and secretion. CLIC5 specifically associates with the cytoskeleton of placenta microvilli. CLIC5 is required for the development and/or maintenance of the proper glomerular endothelial cell and podocyte architecture.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Documents
75789-100.pdf
Datasheet
Technische Daten
Technische Daten
- VE:50 µG
- Validierte Anwendungen:Bioassay
- Purity:> 95% as determined by reducing SDS-PAGE
- Spezies:Human
- Sequence:Met1-Ser251
- Zusammensetzung:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Cat. No.:PRSI91-192
- Ursprung:E. coli
- Molekulargewicht:30.3 kD
- Protein/Peptid-Typ:Rekombinant
- Tag sequence:N-6 His tag
- Lagerbedingungen:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein/Peptid-Name:CLIC5
- Proteinsynonym:Chloride Intracellular Channel Protein 5|CLIC5



