Human recombinant FABP1 (from E. coli)
Catalog # PRSI96-289
Supplier: ProSci Inc.
CAS Number:
Specifications
- VE:200 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Human
- Proteinsynonym:FABPL|FABP1|L-FABP
- Protein/Peptid-Name:FABP1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:15 kDa
- Zusammensetzung:Lyophilized, PBS, pH7.4
- Validierte Anwendungen:Western Blot
Specifications
About this item
Fatty acid-binding protein 1 (FABP1) is also known as Fatty acid-binding protein, liver (FABPL), Liver-type fatty acid-binding protein (L-FABP). FABP1 / FABPL belongs to the calycin superfamily and Fatty-acid binding protein (FABP) family. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 / L-FABP binds free fatty acids and their coenzyme A derivatives, bilirubin, and some other small molecules in the cytoplasm. FABP-1 may be involved in intracellular lipid transport.