Human recombinant CD5 (from HEK293 cells)
Katalognr. PRSI96-118
Anbieter: ProSci Inc.
CAS-Nummer:
Technische Daten
- VE:100 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:HEK293 cells
- Spezies:Human
- Proteinsynonym:CD5|LEU1
- Protein/Peptid-Name:CD5
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:40.5 kDa
- Zusammensetzung:Lyophilized, PBS, pH7.4
- Validierte Anwendungen:Western Blot
Technische Daten
Über diesen Artikel
T-cell surface glycoprotein CD5 is also known as Lymphocyte antigen T1/Leu-1 and LEU1,which is phosphorylated on tyrosine residues by LYN,so CD5 can create binding sites for PTPN6/SHP-1.CD5 may act as a receptor in regulating T-cell proliferation. CD5 is expressed at various developmental and activation stages on human B cells.CD5 is a well established negative regulator of TCR and BCR signalling.CD5-positive cells may also prevent the emergence of autoimmunity by provision of cytokines such as IL-10. Development,selection and function of different B- and T-cell subsets or their preferential survival may be directly or indirectly dependent on different glycan structures associated with CD5 or CD5-like molecules.