Order Entry
Switzerland
ContactUsLinkComponent
Human CDX2 peptide (16 aa near C-terminus)
Catalog # PRSI5595P
Supplier:  ProSci Inc.
CAS Number:  
Human CDX2 peptide (16 aa near C-terminus)
Catalog # PRSI5595P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VE:
    50 µG
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at –20 °C, stable for one year.
  • Aminosäurezahl:
    16 aa near C-terminus
  • Protein/Peptid-Name:
    CDX2 Peptide (16 aa near C-terminus)
  • Validierte Anwendungen:
    Blocking

Specifications

About this item

CDX2 peptide is used for blocking the activity of CDX2 antibody.