Portable/wall group lockout box, Prinzing
N° de catalogue BRDY046134
Fournisseur: Brady
Spécifications
- Typ:Portable/wall group lockout box
- Farbe:Gelb
- B×T×H:354,5×73,4×151 mm
- VE:1
Spécifications
À propos de cet article
This box accommodates 8 to 10 locks (depending on lock size).
- Made of yellow powder-coated metal (mild sheet steel)
- Can be used alone or in combination with Industrial Strength lockout station
Informations de commande: Contents not included.