Anti-PPCDC Rabbit Polyclonal Antibody
Catalog # PRSI30-188
Supplier: ProSci Inc.
Specifications
- Antigen-Name:Phosphopantothenoylcysteine Decarboxylase
- Antigen-Symbol:PPCDC
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human,Dog
- Western blot:Yes
- Form:Lyophilized
- Antigen-Synonyme:coaC|PPC-DC|MDS018
- Immunogen:A synthetic peptide corresponding a region of human PPCDC.
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. PPCDC, one of the last enzymes in this pathway, converts phosphopantothenoylcysteine to 4-prime-phosphopantetheine.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PPCDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human