Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
Switzerland
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
ContactUsLinkComponent
Heim
Standards
Organische Standards
Drug and metabolite standards, desipramine hydrochloride, LoGiCal®
Drug and metabolite standards, desipramine hydrochloride, LoGiCal®
PROOLGCFOR0589.00
:
LGC Standards PROMOCHEM
:
Drug and metabolite standards, desipramine hydrochloride, LoGiCal®
PROOLGCFOR0589.00
:
LGC Standards PROMOCHEM
:
VE:
10 mg
Normen:
Reference standard
CAS Nr.:
58-28-6
Bezeichnung:
Desipramine hydrochloride
Formel:
C₁₈H₂₃ClN₂
Bezeichnung:
Desipramine hydrochloride