Organic reference standard, 4-methyl-1-(3-methylpyridin-2-yl)-2-phenylpiperazine
PROOLGCFOR1005.01
:
- VE:10 mg
- Normen:Reference standard
- CAS Nr.:191546-94-8
- Bezeichnung:4-Methyl-1-(3-methylpyridin-2-yl)-2-phenylpiperazine
- Formel:C₁₇H₂₁N₃
- Bezeichnung:4-Methyl-1-(3-methylpyridin-2-yl)-2-phenylpiperazine