Anti-SLC7A3 Rabbit Polyclonal Antibody
Katalognr. BSBTA09720-1
Anbieter: Boster Bio
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:Cationic amino acid transporter 3
- Antigen-Symbol:SLC7A3
- Klonalität:Polyklonal
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Technische Daten
Über diesen Artikel
Rabbit IgG polyclonal antibody for Cationic amino acid transporter 3(SLC7A3) detection. Tested with WB in Human.
Type: Primary
Antigen: SLC7A3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human