Anti-Follistatin Rabbit Polyclonal Antibody
BSBTA00972-1
- Antikörper-Typ:Primär
- Antigen-Name:Follistatin
- Klonalität:Polyklonal
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Größe:100 µg
- VE:100 µG
Rabbit IgG polyclonal antibody for Follistatin detection. Tested with WB, ELISA(Cap) in Human;Mouse;Rat.
Type: Primary
Antigen: Follistatin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat