Anti-Fabp2 Rabbit Polyclonal Antibody
100 µg
BSBTA02378
Rabbit IgG polyclonal antibody for Fatty acid-binding protein, intestinal(Fabp2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: Fabp2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
- Größe:100 µg
- ImmunoChemie:Yes
- Western blot:Yes
- Cat. No.:BSBTA02378
- Antigen-Name:Fatty acid-binding protein, intestinal
- Antikörper-Typ:Primär
- Reaktivität:Human|Rat|Mouse
- Antigen-Symbol:Fabp2
- Klonalität:Polyklonal
- Wirt:Rabbit



