Order Entry
Switzerland
ContactUsLinkComponent
Impurity and API standard, cytidine
Catalog # PROOMM1316.01
CAS Number:  
Impurity and API standard, cytidine
Catalog # PROOMM1316.01
CAS Number:  

Specifications

  • VE:
    100 mg
  • Normen:
    Reference standards
  • CAS Nr.:
    65-46-3
  • Bezeichnung:
    Cytidine
  • Formel:
    C₉H₁₃N₃O₅
  • Bezeichnung:
    Cytidine

Specifications

About this item