Order Entry
Switzerland
ContactUsLinkComponent
Impurity and API standard, phenylazosalicylic acid
Impurity and API standard, phenylazosalicylic acid
Catalog # PROOMM0217.12
CAS Number:  
Impurity and API standard, phenylazosalicylic acid
Catalog # PROOMM0217.12
CAS Number:  

Specifications

  • VE:
    50 mg
  • Normen:
    Reference standards
  • CAS Nr.:
    3147-53-3
  • Bezeichnung:
    2-Hydroxy-5-(phenyldiazenyl)benzoic acid
  • Formel:
    C₁₃H₁₀N₂O₃
  • Bezeichnung:
    2-Hydroxy-5-(phenyldiazenyl)benzoic Acid (Phenylazosalicylic Acid)

Specifications

About this item