ContactUsLinkComponent
Organic reference standards, Benserazide hydrochloride assay standard, BP
Catalog # PROOBP685
CAS Number:  
Organic reference standards, Benserazide hydrochloride assay standard, BP
Catalog # PROOBP685
CAS Number:  
Specifications
  • VE:
    100 mg
  • Normen:
    Reference standard
  • CAS Nr.:
    14919-77-8
  • Bezeichnung:
    Benserazide hydrochloride assay standard
  • Formel:
    C₁₀H₁₅N₃O₅·ClH
  • Bezeichnung:
    Benserazide hydrochloride assay standard