Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organische Standards
Organic reference standards, Adapalene (assay standard), BP
Organic reference standards, Adapalene (assay standard), BP
Organic reference standards, Adapalene (assay standard), BP
PROOBP1120
:
LGC Standards PROMOCHEM
:
Organic reference standards, Adapalene (assay standard), BP
PROOBP1120
:
LGC Standards PROMOCHEM
:
VE:
100 mg
Normen:
Reference Standard
CAS Nr.:
106685-40-9
Bezeichnung:
Bp-1120, adapalene (assay standard)
Formel:
C₂₈H₂₈O₃
Bezeichnung:
Adapalene