Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standards, Chlorhexidine acetate assay standard, BP
Organic reference standards, Chlorhexidine acetate assay standard, BP
Catalog # PROOBP068
CAS Number:  
Organic reference standards, Chlorhexidine acetate assay standard, BP
Catalog # PROOBP068
CAS Number:  

Specifications

  • VE:
    100 mg
  • Normen:
    Reference Standard
  • CAS Nr.:
    57-95-1
  • Bezeichnung:
    Chlorhexidine acetate assay standard, BP 068
  • Formel:
    C₁₄H₉Cl₅
  • Bezeichnung:
    Chlorhexidine acetate assay standard

Specifications

About this item