Order Entry
Switzerland
ContactUsLinkComponent
Anti-beta subunit Cholera Toxin Rabbit Polyclonal Antibody
Katalognr. BIRBORB159212-100
Anbieter:  Biorbyt
Anti-beta subunit Cholera Toxin Rabbit Polyclonal Antibody
Katalognr. BIRBORB159212-100
Anbieter:  Biorbyt

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    beta subunit Cholera Toxin
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Bacteria
  • Lagertemperatur:
    Store at –20 °C for 12 months
  • Immunogen:
    KLH conjugated synthetic peptide derived from beta subunit Cholera Toxin
  • VE:
    100 µl

Technische Daten

Über diesen Artikel

Type: Primary
Antigen: beta subunit Cholera Toxin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacterial