Anti-LOXL2 Rabbit Polyclonal Antibody
Catalog # BIRBORB184846-200
Anbieter: Biorbyt
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Symbol:LOXL2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Lagertemperatur:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at –20 °C, avoid freeze/thaw cycles
- Immunogen:KLH conjugated synthetic peptide derived from human LOXL2
- VE:200 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: LOXL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat