Order Entry
Switzerland
ContactUsLinkComponent
Anti-NFKBIA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-NFKBIA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
N° de catalogue BOSSBS-5515R-A647
Fournisseur:  Bioss
Anti-NFKBIA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
N° de catalogue BOSSBS-5515R-A647
Fournisseur:  Bioss

Spécifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Nuclear Factor of Kappa Light Polypeptide Gene Enhancer in B-cells Inhibitor, Alpha
  • Antigen-Symbol:
    NFKBIA
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 647
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    4792
  • Antigen-Synonyme:
    MAD3|IkappaBalpha|IkB-alpha|Major histocompatibility complex enhancer-binding protein MAD3|NFKBI|NF-kappa-B inhibitor alpha|I-kappa-B-alpha|IKBA
  • Modification:
    phospho-Tyr305
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Immunogen:
    295-310/317
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Spécifications

À propos de cet article

Inhibits the activity of dimeric NF-kappa-B/REL complexes by trapping REL dimers in the cytoplasm through masking of their nuclear localization signals. On cellular stimulation by immune and proinflammatory responses, becomes phosphorylated promoting ubiquitination and degradation, enabling the dimeric RELA to translocate to the nucleus and activate transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NFKBIA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat