Order Entry
Switzerland
ContactUsLinkComponent
Anti-HTR3B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-HTR3B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-4289R-A647
Supplier:  Bioss
Anti-HTR3B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-4289R-A647
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    5-Hydroxytryptamine (serotonin) Receptor 3B
  • Antigen-Symbol:
    HTR3B
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 647
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    9177
  • Antigen-Synonyme:
    5-HT3B
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Immunogen:
    130-180/441
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Specifications

About this item

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarizing responses. It is a cation-specific, but otherwise relatively nonselective, ion channel.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HTR3B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat