Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD19 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalognr. BOSSBS-10358R-FITC
Anbieter:  Bioss
Anti-CD19 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalognr. BOSSBS-10358R-FITC
Anbieter:  Bioss

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    CD19 molecule
  • Antigen-Symbol:
    CD19
  • Klonalität:
    Polyklonal
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Dog
  • Kreuzadsorption:
    No
  • Form:
    liquid
  • Antigen-Synonyme:
    B-lymphocyte antigen CD19. B4|CVID3|AW495831|mouse CD19|Cluster of Differentiation 19
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Konzentration:
    1 μg/μl
  • Transporttemperatur:
    4°C
  • Immunogen:
    170-220/611
  • Reinigung:
    Purified by Protein A
  • VE:
    100 µl

Technische Daten

Über diesen Artikel

9 is a transmembrane glycoprotein that is selectively expressed on the cell surface of B-lymphocytes,where it activates intracellular signaling cascades involving both Ras and phosphatidylinositol 3-kinase pathways.Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. This gene encodes a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD19
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Dog