Anti-NFKBIA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-1287R-FITC
: Bioss
- Antikörper-Typ:Primär
- Antigen-Name:Nuclear Factor of Kappa Light Polypeptide Gene Enhancer in B-cells Inhibitor, Alpha
- Antigen-Symbol:NFKBIA
- Klonalität:Polyklonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Kreuzadsorption:No
- Form:liquid
- Gen-ID:18035
- Antigen-Synonyme:MAD3|IkappaBalpha|IkB-alpha|Major histocompatibility complex enhancer-binding protein MAD3|NFKBI|NF-kappa-B inhibitor alpha|I-kappa-B-alpha|IKBA
- Lagerungspuffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Lagertemperatur:Store at 4°C for 12 months
- Konzentration:1 μg/μl
- Transporttemperatur:4°C
- Immunogen:5-34/314
- Reinigung:Purified by Protein A
- VE:100 µl
Inhibits the activity of dimeric NF-kappa-B/REL complexes by trapping REL dimers in the cytoplasm through masking of their nuclear localization signals. On cellular stimulation by immune and proinflammatory responses, becomes phosphorylated promoting ubiquitination and degradation, enabling the dimeric RELA to translocate to the nucleus and activate transcription.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NFKBIA/IKB alpha
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat