ContactUsLinkComponent
Tubes and bottles for Beckman rotors
  BECK357005
Tubes and bottles for Beckman rotors
  BECK357005
  • Bezeichnung:
    Tube, PP, translucent, with snap-on cap, 50 ml, 29×104 mm
  • Seq:
    08
  • VE:
    25
An assortment of tubes and bottles for use with Beckman rotors.
 : Tubes and bottles are not supplied with lids. Lids must be ordered separately.