Anti-Nrf3 Rabbit Polyclonal Antibody [clone: D530]
BWRLBS9275
- Antikörper-Typ:Primär
- Antigen-Symbol:Nrf3
- Klonalität:Polyklonal
- Klon:D530
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Synthetic peptide, corresponding to amino acids 500-520 of Human Nrf3.
Nrf3 (D530) pAb detects endogenous levels of Nrf3 protein.
Type: Primary
Antigen: Nrf3
Clonality: Polyclonal
Clone: D530
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human