Anti-p-Catenin-beta Rabbit Polyclonal Antibody [clone: T41/S45]
BWRLBS5057
- Antikörper-Typ:Primär
- Antigen-Symbol:p-Catenin- beta
- Klonalität:Polyklonal
- Klon:T41/S45
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
A phospho specific peptide corresponding to residues surrounding threonine 41 and 45 of Human Catenin-beta.
p-beta-catenin (T41/S45) pAb detects endogenous levels of beta-catenin protein only when phosphorylated at Thr41/S45.
Type: Primary
Antigen: p-Catenin-beta
Clonality: Polyclonal
Clone: T41/S45
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat