Anti-p-Csk Rabbit Polyclonal Antibody [clone: S364]
Katalognr. BWRLBS4673
Anbieter: Bioworld Technology
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Symbol:p-Csk
- Klonalität:Polyklonal
- Klon:S364
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Technische Daten
Über diesen Artikel
Synthetic phosphopeptide derived from human Csk around the phosphorylation site of Serine 364.
p-Csk (S364) pAb detects endogenous levels of Csk protein when phosphorylated at Ser364.
Type: Primary
Antigen: p-Csk
Clonality: Polyclonal
Clone: S364
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat