Anti-p-CASP8 Rabbit Polyclonal Antibody [clone: Y380]
Katalognr. BWRLBS4662
Anbieter: Bioworld Technology
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Symbol:p-CASP8
- Klonalität:Polyklonal
- Klon:Y380
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Technische Daten
Über diesen Artikel
Synthetic phosphopeptide derived from human Caspase-8 around the phosphorylation site of Tyrosine 380.
p-Caspase 8 (Y380) pAb detects endogenous levels of Caspase 8 protein only when phosphorylated at Tyr380.
Type: Primary
Antigen: p-CASP8
Clonality: Polyclonal
Clone: Y380
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat