Anti-p-MDM2 Rabbit Polyclonal Antibody [clone: S166]
Catalog # BWRLBS4273
Supplier: Bioworld Technology
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:p-MDM2
- Klonalität:Polyklonal
- Klon:S166
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human MDM2 around the phosphorylation site of Serine 166.
p-MDM2 (S166) pAb detects endogenous levels of MDM2 protein only when phosphorylated at Ser166
Type: Primary
Antigen: p-MDM2
Clonality: Polyclonal
Clone: S166
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat