Teepol, multi purpose detergent
Catalog # DIVYF7316
Supplier: Diversey
Specifications
- Bezeichnung:Teepol, multi-purpose detergent
- Verpackt:5 litre container
- VE:2 Can
Specifications
About this item
Teepol is a multi-purpose detergent for cleaning all washable surfaces and recommended for the food industry.
- Used as a wetting agent and aggressively scented
- Facilitating the elimination of the stubborn dirt
- Effective emulsifying power, allowing a rapid dispersion of fats and oils
- Excellent degreaser
It is very effective in industrial or agricultural applications.