Order Entry
Switzerland
ContactUsLinkComponent
Anti-PD-L1/B7-H1 Rabbit Polyclonal Antibody
Katalognr. BSBTA00109
Supplier:  Boster Bio
undefined
Anti-PD-L1/B7-H1 Rabbit Polyclonal Antibody
Katalognr. BSBTA00109
Supplier:  Boster Bio

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    PD-L1/B7-H1
  • Klonalität:
    Polyklonal
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Größe:
    100 µg
  • VE:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Programmed cell death 1 ligand 1(CD274) detection. Tested with WB in Human.

Type: Primary
Antigen: PD-L1/B7-H1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human