Phosphate buffered saline, PBS-TWEEN® tablets, Millipore®
Catalog # 524653-1
Supplier: Merck Millipore (Oncogene)
CAS Number:
Specifications
- Packung:Glass bottle
- VE:10 Tab.
- Bezeichnung:PBS-TWEEN® tablets, pH 7,4
Specifications
About this item
Developed for use in immunoassay applications.
One tablet dissolved in 1 litre of deionised water yields 140 mM NaCl, 3 mM KCl, 0,05% TWEEN 20 detergent, 10 mM phosphate buffer with a pH of 7,4 at 25 °C.
Can be stored at 15 to 30 °C.