Human alpha 1 Fetoprotein ELISA kit
BIRBORB78656-96
: Biorbyt
:
- VE:96
- Reaktivität der primären Antikörper:Human
- Bezeichnung:Human Alpha 1 Fetoprotein ELISA Kit
Human Alpha 1 Fetoprotein (AFP) ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of AFP in biological samples such as serum, plasma or cell culture supernatants.
: Kit includes pre-coated 96-well strip microplate, Wash Buffer Concentrate 20X, 25 ml; standards, 2 vials; Assay Diluent A, 30 ml; Assay Diluent B 5X, 15 ml; detection antibody, 2 vials; HRP-streptavidin concentrate, 200 µl; TMB One-Step Substrate Reagent, 12 ml; and Stop Solution, 8 ml.