AuftragsverfolgungKontakt
Human galectin-9 ELISA kit | Avantor
Human galectin-9 ELISA kit
Human Galectin-9 ELISA Kit, 96
Katalognr. BIRBORB138106-96
Marketsource
Anbieter:   Biorbyt
Human galectin-9 ELISA kit
Katalognr. BIRBORB138106-96
Anbieter:  Biorbyt
Marketsource
CAS Nummer:  
Human galectin-9 ELISA kit
Human galectin-9 ELISA kit
Human Galectin-9 ELISA Kit, 96
CAS Nummer:  
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 96 ST
Product Details & Documents
Human galectin-9 ELISA kit for the quantitative detection of human galectin-9 in cell culture supernates, serum and plasma (heparin, EDTA).
  • No detectable cross-reactivity with other relevant proteins
Galectin-9, also called HUAT or LGALS9A is a protein that in humans is encoded by the LGALS9 gene. This gene is mapped to 17q11.2. The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkins disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. The protein has N- and C- terminal carbohydrate-binding domains connected by a link peptide.
Dokumente
Technische Daten
Technische Daten
  • VE:
    96
  • Bezeichnung:
    Human Galectin-9 ELISA Kit
  • Cat. No.:
    BIRBORB138106-96
  • Empfindlichkeit:
    <10 pg/ml
  • Reaktivität der primären Antikörper:
    Human
  • Erfassungsbereich:
    93,8 - 6000 pg/ml