Mouse CXCL13 ELISA kit
Katalognr. BIRBORB106616-96
Anbieter: Biorbyt
CAS-Nummer:
Technische Daten
- VE:96
- Reaktivität der primären Antikörper:Mouse
- Bezeichnung:Mouse CXCL13 ELISA Kit
- Erfassungsbereich:15,6 - 1000 pg/ml
- Empfindlichkeit:<10 pg/ml
Technische Daten
Über diesen Artikel
Mouse CXCL13 ELISA Kit for the quantitative determination of mouse Chemokine (C-X-C motif) 13 (CXCL13).
CXCL13, also known as B lymphocyte chemoattractant (BLC), is a protein that in humans is encoded by the CXCL13 gene. CXCL13 is a small cytokine belonging to the CXC chemokine family, and it was located on sequence maps to 4q21, near the genes encoding most CXC chemokines. In T-lymphocytes, CXCL13 expression is thought to reflect a germinal centre origin of the T-cell. Hence, expression of CXCL13 in T-cell lymphomas, such as Angioimmunoblastic T-cell Lymphoma, is thought to reflect a germinal center origin of the neoplastic T-cells.