Human B7-1 ELISA kit
Katalognr. BIRBORB106611-96
Anbieter: Biorbyt
CAS-Nummer:
Technische Daten
- VE:96
- Reaktivität der primären Antikörper:Human
- Bezeichnung:Human B7-1 ELISA kit, 96-well
- Erfassungsbereich:62,5 - 4000 pg/ml
- Empfindlichkeit:<10 pg/ml
Technische Daten
Über diesen Artikel
B7-1 also known as protein CD80 is a molecule found on activated B cells and monocytes which provides a costimulatory signal necessary for T cell activation and survival.
The cDNA for B7-1 predicts a type I membrane protein. The protein is predicted to contain two extracellular domains structurally similar to those of Ig, a hydrophobic transmembrane region, and a short cytoplasmic domain.
The CD80 and CD86 genes encode B7-1 and B7-2, respectively, which are structurally similar members of the immunoglobulin superfamily expressed on a variety of hematopoietic cell types. B7-1 and B7-2 provide a costimulatory signal to T cells by interacting with CD28 and CTLA4.