Human recombinant Cystatin C (from E. coli)
: Biorbyt
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Human
- Lagerbedingungen:Store lyophilized protein at C20 °C. Lyophilized protein remains stable until the expiry date when stored at C20 °C. Aliquot reconstituted protein to avoid repeated freezing/thawing cycles and store at C80C for long term storage. Reconstituted protein can be stored at 4C for limited period of time; it does not show any change after two weeks at 4C.
- Proteinsynonym:AD8
- Protein/Peptid-Name:Cystatin C
- Purity:Greater than 95% as determined by SDS PAGE.
- Solubility:Add deionized water to prepare working stock solution of approximately 0.5mg/mL and let the lyophilized pellet dissolve completely. Product is not sterile! Please filter the product by an appropriate sterile filter before using it in the cell culture.
- Zusammensetzung:Lyophilized from from 0.5mg/mL in 20 mM Tris buffer and 50mM NaCl pH-7.5.
- Validierte Anwendungen:SDS-PAGE
Frequently Bought Together