Human DDIT3 (from E. coli)
Catalog # BIRBORB81067-5
Supplier: Biorbyt
CAS Number:
Specifications
- VE:5 µG
- Ursprung:E. coli
- Spezies:Human
- Lagerbedingungen:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Proteinsynonym:C
- Protein/Peptid-Name:DDIT3
- Zusammensetzung:DDIT3 protein at concentration 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
- Validierte Anwendungen:SDS-PAGE